Få mere at vide
3-Methyl-d3-indole, TRC
Artikelnummer. 30473575
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30473575 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473575 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31347225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-d3-indole
CAS 111399-60-1
Molekylær formel C9 2H3 H6 N
Mængde 25 mg
Synonym 1H-Indole','3-(methyl-d3)-, 3-(Methyl-d3)-1H-indole, 3-Methyl-1H-indole-d3, 3-Methylindole-D3 (methyl-D3), Scatole-d3, Skatol-d3, β-Methylindole-d3
InChI formel InChI=1S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10H,1H3/i1D3
SMIL [2H]C([2H])([2H])c1c[nH]c2ccccc12
IUPAC navn 3-(trideuteriomethyl)-1H-indole
Molekylvægt (g/mol) 134.19
Formel vægt 134.0923