Få mere at vide
3-Methyl-d3-4-nitrophenol-2,5,6-d3, TRC
Artikelnummer. 30464570
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30464570 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464570 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49375625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-4-nitrophenol methyl,ring-d6
CAS 1219803-89-0
Molekylær formel C7 2H6 H N O3
Mængde 25 mg
Synonym 3-Methyl-4-nitrophenol-D6, 3-(Methyl-d3)-4-nitrophenol-2,5,6-d3, 2-Nitro-5-hydroxytoluene-d6, 4-Nitro-m-cresol-d6, 5-Hydroxy-2-nitrotoluene-α,α,α,3,4,6-d6, 5-Methyl-4-nitrophenol-d6
InChI formel InChI=1S/C7H7NO3/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4,9H,1H3/i1D3,2D,3D,4D
SMIL [2H]c1c([2H])c(c(c([2H])c1O)C([2H])([2H])[2H])[N+](=O)[O-]
IUPAC navn 2,3,6-trideuterio-4-nitro-5-(trideuteriomethyl)phenol
Molekylvægt (g/mol) 159.17
Formel vægt 159.0803