Få mere at vide
3-Methyl Adenine-d3, TRC
Artikelnummer. 30477294
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477294 1 mg
30464524 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477294 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2759051MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110953-39-4
Molekylær formel C6 2H3 H4 N5
Molekylvægt (g/mol) 152.17
Synonym 3-Methyl Adenine D3
IUPAC navn 3-(trideuteriomethyl)purin-6-amine
SMIL [2H]C([2H])([2H])n1cnc(N)c2ncnc12
Tekniske data
Kemisk navn eller materiale 3-Methyl-d3-adenine
Mængde 1 mg
InChI formel InChI=1S/C6H7N5/c1-11-3-10-5(7)4-6(11)9-2-8-4/h2-3H,7H2,1H3/i1D3
Formel vægt 152.089