missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(pentafluorothio)benzaldehyde, 97%, Thermo Scientific™

Catalog Number 15436627
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1240257-01-5
C8H7F5OS
246.195
MFCD16652480
MBCKIHKPQIVSBW-UHFFFAOYSA-N
3-methyl-5-pentafluorosulfur benzaldehyde, 3-methyl-5-pentafluoro-??-sulfanyl benzaldehyde, 3-methyl-5-pentafluorothio benzaldehyde
66523591
3-methyl-5-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
CC1=CC(=CC(=C1)C=O)S(F)(F)(F)(F)F
This item is not returnable. View return policy
1240257-01-5
C8H7F5OS
246.195
MFCD16652480
MBCKIHKPQIVSBW-UHFFFAOYSA-N
3-methyl-5-pentafluorosulfur benzaldehyde, 3-methyl-5-pentafluoro-??-sulfanyl benzaldehyde, 3-methyl-5-pentafluorothio benzaldehyde
66523591
3-methyl-5-(pentafluoro-$l^{6}-sulfanyl)benzaldehyde
CC1=CC(=CC(=C1)C=O)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 246.19
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(pentafluorothio)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'