Få mere at vide
3-Methyl-5-[2,6,6-trimethyl-1-(cyclohexen-d5)-1-yl]-penta-2,4-dienenitrile, TRC
Artikelnummer. 30474359
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30474359 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474359 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33096750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-5-[2,6,6-trimethyl-1-(cyclohexen-d5)-1-yl]-penta-2,
CAS 1185235-72-6
Molekylær formel C15 2H5 H16 N
Mængde 50 mg
Synonym 3-Methyl-5-[2,6,6-trimethyl-1-(cyclohexen-d5)-1-yl]-penta-2,4-dienenitrile, 5-[3,3-Dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]-3-methylpenta-2,4-dienenitrile, β-Ionylideneacetonitrile-d5, 3-Methyl-5-(2,6,6-trimethyl-1-cyclohexen-1-yl)penta-2,4-dienenitrile-D5 (6,6-dimethyl-2-(methyl-D3)-cyclohexen-3,3-D2)
InChI formel InChI=1S/C15H21N/c1-12(9-11-16)7-8-14-13(2)6-5-10-15(14,3)4/h7-9H,5-6,10H2,1-4H3/b8-7+,12-9+/i2D3,6D2
SMIL [2H]C([2H])([2H])C1=C(\C=C\C(=C\C#N)\C)C(C)(C)CCC1([2H])[2H]
IUPAC navn (2E,4E)-5-[3,3-dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]-3-methylpenta-2,4-dienenitrile
Molekylvægt (g/mol) 220.36
Formel vægt 220.1988