Få mere at vide
3-Methyl-4-nitroimino-tetrahydro-1,3,5-oxadiazine, TRC
Artikelnummer. 30006629
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006629 5 g
30006246 1 g
30004533 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006629 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3238505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 153719-38-1
Molekylær formel C4 H8 N4 O3
Molekylvægt (g/mol) 160.13
Synonym 3-Methyl-4-nitroimino-tetrahydro-1,3,5-oxadiazine
IUPAC navn N-(3-methyl-1,3,5-oxadiazinan-4-ylidene)nitramide
SMIL CN1COCNC1=N[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 3-Methyl-4-nitroimino-tetrahydro-1,3,5-oxadiazine
Mængde 5 g
InChI formel InChI=1S/C4H8N4O3/c1-7-3-11-2-5-4(7)6-8(9)10/h2-3H2,1H3,(H,5,6)
Formel vægt 160.0596