Få mere at vide
3-Methyl-3-buten-2-one (Stabilized in HQ) (~90%), TRC
Artikelnummer. 30482925
Skift visning
Mængde:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482925 1 mL
30466508 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482925 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3328551ML
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 814-78-8
Molekylær formel C5H8O
Molekylvægt (g/mol) 84.12
Synonym 2-Methyl-1-buten-3-one, 3-Methylene-2-butanone, Isopropenyl Methyl Ketone, Methyl Isopropenyl Ketone, NSC 24150, Propen-2-yl Methyl Ketone
IUPAC navn 3-methylbut-3-en-2-one
SMIL CC(=C)C(=O)C
Tekniske data
Kemisk navn eller materiale 3-Methyl-3-buten-2-one
Mængde 1 mL
InChI formel InChI=1S/C5H8O/c1-4(2)5(3)6/h1H2,2-3H3
Formel vægt 84.0575