Få mere at vide
3-Methyl-2-buten-1-ol-d6 (d5 Major)(Contain ~3% d0), TRC
Artikelnummer. 30464980
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30464980 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464980 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29439725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-2-buten-1-ol-d6
CAS 53439-16-0
Molekylær formel C5 2H6 H4 O
Mængde 25 mg
Synonym 2-Buten-4,4,4-d3-1-ol, 3-(methyl-d3)-, 3-(Methyl-d3)-2-buten-4,4,4-d3-1-ol, 3-Methyl-2-buten-1-ol-d6, 3-Methyl-2-buten-1-ol-D6 (dimethyl-D6), Prenol-d6, Prenyl alcohol D6, γ,γ-Di(methyl-d3)allyl alcohol
InChI formel InChI=1S/C5H10O/c1-5(2)3-4-6/h3,6H,4H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])C(=CCO)C([2H])([2H])[2H]
IUPAC navn 4,4,4-trideuterio-3-(trideuteriomethyl)but-2-en-1-ol
Molekylvægt (g/mol) 92.17
Formel vægt 92.1108