Få mere at vide
3-Methyl-1H-Indole-d8, TRC
Artikelnummer. 30477216
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30477216 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477216 Leverandør Toronto Research Chemicals Leverandørnr. TRCM313473100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-1H-Indole-d8
CAS 697807-03-7
Molekylær formel C9 2H8 H N
Mængde 100 mg
Synonym 1H-Indole-2,4,5,6,7-d5','3-(methyl-d3)-, 3-(Methyl-d3)-1H-indole-2,4,5,6,7-d5, 3-Methyl-1H-indole-d8, 3-Methylindole-D8, Scatole-d8, Skatol-d8, β-Methylindole-d8
InChI formel InChI=1S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10H,1H3/i1D3,2D,3D,4D,5D,6D
SMIL [2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1C([2H])([2H])[2H]
IUPAC navn 2,4,5,6,7-pentadeuterio-3-(trideuteriomethyl)-1H-indole
Molekylvægt (g/mol) 139.22
Formel vægt 139.1237