Få mere at vide
3-Methyl-1-butyl-1,1-d2 Alcohol, CDN
Artikelnummer. 30310508
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310508 0.25 g -
30318399 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310508 Leverandør CDN Leverandørnr. CDND62330.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-1-butanol-1,1-D2
CAS 70907-83-4
Molekylær formel C5 D2 H10 O
Mængde 0.25 g
Synonym 1-Butan-1,1-d2-ol, 3-methyl- (9 CI, ACI), 3-Methyl-1-butan-1,1-d2-ol (ACI), 3-Methyl-1-butyl-1,1-d2 alcohol, 3-Methyl-1-butanol-1,1-d2, 3-Methyl-1-butanol-1,1-D2
InChI formel InChI=1 S/C5H12O/c1-5(2)3-4-6/h5-6 H,3-4H2,1-2H3/i4D2
SMIL [2 H]C([2 H])(O)CC(C)C
IUPAC navn 1,1-dideuterio-3-methylbutan-1-ol
Molekylvægt (g/mol) 90.1605
Formel vægt 90.1014 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity