Få mere at vide
3-Methyl-1,5-pentanediol Diacrylate (~0.5% 4-Hydroxy Anisole as a stabilizer), TRC
Artikelnummer. 30133320
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133320 1 g
30134589 250 mg
30006604 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133320 Leverandør Toronto Research Chemicals Leverandørnr. TRCM9177801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64194-22-5
Molekylær formel C12H18O4
Molekylvægt (g/mol) 226.27
Synonym 2-Propenoic acid, 3-methyl-1,5-pentanediyl Ester, 3-Methylpentanediol diacrylate, Light Acrylate MPD-A, MPD-A, Photomer 4071F, SR 341, Sartomer SR 341
IUPAC navn (3-methyl-5-prop-2-enoyloxypentyl) prop-2-enoate
SMIL CC(CCOC(C=C)=O)CCOC(C=C)=O
Tekniske data
Kemisk navn eller materiale 3-Methyl-1,5-pentanediol Diacrylate
Mængde 1 g
InChI formel InChI=1S/C12H18O4/c1-4-11(13)15-8-6-10(3)7-9-16-12(14)5-2/h4-5,10H,1-2,6-9H2,3H3
Formel vægt 226.1205