missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Methoxyisobenzofuran-1(3H)-one, TRC

Artikelnummer. 30468613
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
Pakningsstørrelse:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30468613 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30468613 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2632702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Methoxyisobenzofuran-1(3H)-one
CAS 4122-57-0
Molekylær formel C9H8O3
Mængde 2.5 g
Synonym 3-Methoxyphthalide, NSC 408564, 3-Methoxy-1(3H)-isobenzofuranone, 3-Methoxy-2-benzofuran-1(3H)-one, 3-Methoxy-3H-isobenzofuran-1-one
InChI formel InChI=1S/C9H8O3/c1-11-9-7-5-3-2-4-6(7)8(10)12-9/h2-5,9H,1H3
SMIL COC1OC(=O)c2ccccc12
IUPAC navn 3-methoxy-3H-2-benzofuran-1-one
Molekylvægt (g/mol) 164.16
Formel vægt 164.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.