Få mere at vide
3-Methoxy-propionaldehyde (Technical Grade), TRC
Artikelnummer. 30468833
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468833 500 mg
30468479 100 mg
30469249 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30468833 Leverandør Toronto Research Chemicals Leverandørnr. TRCM336193500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2806-84-0
Molekylær formel C4H8O2
Molekylvægt (g/mol) 88.11
IUPAC navn 3-methoxypropanal
SMIL COCCC=O
Tekniske data
Kemisk navn eller materiale 3-Methoxy-propionaldehyde (Technical Grade)
Mængde 500 mg
InChI formel InChI=1S/C4H8O2/c1-6-4-2-3-5/h3H,2,4H2,1H3
Formel vægt 88.0524