Få mere at vide
3-(Methoxy-d3)-1-propanol, TRC
Artikelnummer. 30465772
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30465772 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465772 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26656225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(Methoxy-d3)-1-propanol
CAS 86013-00-5
Molekylær formel C4H7D3O2
Mængde 25 mg
InChI formel InChI=1S/C4H10O2/c1-6-4-2-3-5/h5H,2-4H2,1H3/i1D3
SMIL OCCCOC([2H])([2H])[2H]
IUPAC navn 3-(methoxy-d3)propan-1-ol
Molekylvægt (g/mol) 93.14
Formel vægt 93.09