Learn More
Alfa Aesar™ 3-Methoxy-4-(trifluoromethyl)benzyl bromide, 97%
Brand: Alfa Aesar™ H33880.MD
Additional Details : Weight : 0.00025kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Quantity | 250mg |
---|
Chemical Identifiers
853367-87-0 | |
269.061 | |
MUVUCILNTIWNQL-UHFFFAOYSA-N | |
66523605 | |
COC1=C(C=CC(=C1)CBr)C(F)(F)F |
C9H8BrF3O | |
MFCD18398596 | |
3-methoxy-4-trifluoromethyl benzyl bromide, 4-bromomethyl-2-methoxy-1-trifluoromethyl benzene | |
4-(bromomethyl)-2-methoxy-1-(trifluoromethyl)benzene |
Specifications
3-Methoxy-4-(trifluoromethyl)benzyl bromide | |
853367-87-0 | |
MFCD18398596 | |
3-methoxy-4-trifluoromethyl benzyl bromide, 4-bromomethyl-2-methoxy-1-trifluoromethyl benzene | |
COC1=C(C=CC(=C1)CBr)C(F)(F)F | |
269.061 | |
269.06 | |
97% |
250mg | |
C9H8BrF3O | |
UN3261 | |
MUVUCILNTIWNQL-UHFFFAOYSA-N | |
4-(bromomethyl)-2-methoxy-1-(trifluoromethyl)benzene | |
66523605 | |
Solid |