missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-methylpyridine, 97%, Acros Organics™

Brand:  Acros Organics 453720010

Additional Details : CAS Number : 26395-26-6 Weight : 0.01000kg


Quantity 1g

 View more versions of this product

Product Code. 15296038

  • 694.34 DKK / 1g


Estimated Shipment date
from Supplier 09-06-2020
Add to basket


Specifications

Chemical Identifiers

26395-26-6
123.16
3-methoxy-2-picoline, 3-methoxy-2-methyl-pyridine, pyridine,3-methoxy-2-methyl, pubchem6637, 2-methyl-3-methoxypyridine
3-methoxy-2-methylpyridine
C7H9NO
QRCUQLANPHYVEH-UHFFFAOYSA-N
3015270
CC1=C(C=CC=N1)OC

Specifications

3-Methoxy-2-methylpyridine
Authentic
170.0°C to 175.0°C
26395-26-6
C7H9NO
QRCUQLANPHYVEH-UHFFFAOYSA-N
3-methoxy-2-methylpyridine
3015270
97%
61°C
Glass Bottle
1g
96% min. (GC)
3-methoxy-2-picoline, 3-methoxy-2-methyl-pyridine, pyridine,3-methoxy-2-methyl, pubchem6637, 2-methyl-3-methoxypyridine
CC1=C(C=CC=N1)OC
123.16
123.16
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methoxy-2-methylpyridine, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents