Få mere at vide
3-Methoxy-2',4',6',4-tetrahydroxychalcone (>80%), TRC
Artikelnummer. 30465410
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465410 250 mg
30467267 50 mg
30471750 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30465410 Leverandør Toronto Research Chemicals Leverandørnr. TRCM269613250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52218-19-6
Molekylær formel C16H14O6
Molekylvægt (g/mol) 302.28
Synonym 2',4,4',6'-Tetrahydroxy-3-methoxychalcone, 3-(4-Hydroxy-3-methoxyphenyl)-1-(2,4,6-trihydroxyphenyl)-2-propen-1-one
IUPAC navn 3-(4-hydroxy-3-methoxyphenyl)-1-(2,4,6-trihydroxyphenyl)prop-2-en-1-one
SMIL COc1cc(C=CC(=O)c2c(O)cc(O)cc2O)ccc1O
Tekniske data
Kemisk navn eller materiale 3-Methoxy-2',4',6',4-tetrahydroxychalcone
Mængde 250 mg
InChI formel InChI=1S/C16H14O6/c1-22-15-6-9(2-4-11(15)18)3-5-12(19)16-13(20)7-10(17)8-14(16)21/h2-8,17-18,20-21H,1H3
Formel vægt 302.079