Få mere at vide
3-Methanethiosulfonyl-N,N-dimethylpropionamide, TRC
Artikelnummer. 30473555
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473555 25 mg
30479191 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473555 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25900025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methanethiosulfonyl-N,N-dimethylpropionamide
CAS 359436-82-1
Molekylær formel C6 H13 N O3 S2
Mængde 25 mg
InChI formel InChI=1S/C6H13NO3S2/c1-7(2)6(8)4-5-11-12(3,9)10/h4-5H2,1-3H3
SMIL CN(C)C(=O)CCSS(=O)(=O)C
IUPAC navn N,N-dimethyl-3-methylsulfonylsulfanylpropanamide
Molekylvægt (g/mol) 211.3
Formel vægt 211.0337