Learn More
3-Mercapto-1,2-propanediol, Sigma-Aldrich™
Product Code. 15824811
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity Unit Size
15824811 50 mL 50mL
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15824811 Supplier Sigma Aldrich Supplier No. 8417050050
 Restricted Item
Forventet forsendelse: 24-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 96-27-5
Melting Point <-20°C
Density 1.247 g/cm3 at 20°C
MDL Number MFCD00004879
Quantity 50 mL
Synonym 3-Mercapto-1,2-propanediol, 1-Thioglycerol, 3-Mercapto-1,2-propanediol
Molecular Weight (g/mol) 108.16
Grade Research