Learn More
3-Maleimidobenzonitrile, 97%, Thermo Scientific™
Catalog Number 15424758
Click to view available options
Quantity:
1 g
5 g
303770-14-1
C11H6N2O2
198.18
MFCD00176056
DZBWWFCXELLNDA-UHFFFAOYSA-N
3-2,5-dioxopyrrol-1-yl benzonitrile, cyto9h4, 3-maleimidobenzonitrile, maybridge3_005826, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzonitrile
2732154
O=C1C=CC(=O)N1C1=CC=CC(=C1)C#N
303770-14-1
C11H6N2O2
198.18
MFCD00176056
DZBWWFCXELLNDA-UHFFFAOYSA-N
3-2,5-dioxopyrrol-1-yl benzonitrile, cyto9h4, 3-maleimidobenzonitrile, maybridge3_005826, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzonitrile
2732154
O=C1C=CC(=O)N1C1=CC=CC(=C1)C#N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
IUPAC Name 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzonitrile
Formula Weight 198.18
Percent Purity 97%
Chemical Name or Material 3-Maleimidobenzonitrile