Learn More
3-Isocyanato-5-methyl-2-phenylfuran, 97%, Thermo Scientific™
Catalog Number p-3485491
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
898289-18-4
C12H9NO2
199.209
MFCD09025851
GDLFJANXKGIZKA-UHFFFAOYSA-N
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
18525836
3-isocyanato-5-methyl-2-phenylfuran
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
898289-18-4
C12H9NO2
199.209
MFCD09025851
GDLFJANXKGIZKA-UHFFFAOYSA-N
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
18525836
3-isocyanato-5-methyl-2-phenylfuran
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-Isocyanato-5-methyl-2-phenylfuran