missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-nitroanisole, 97%, Thermo Scientific Chemicals

Catalog Number p-4510565
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
214279-40-0
C7H6INO3
279.03
MFCD01320682
NLTAHTYBZZSCIT-UHFFFAOYSA-N
3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
2733456
2-iodo-4-methoxy-1-nitrobenzene
COC1=CC=C(C(I)=C1)[N+]([O-])=O
This item is not returnable. View return policy
214279-40-0
C7H6INO3
279.03
MFCD01320682
NLTAHTYBZZSCIT-UHFFFAOYSA-N
3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
2733456
2-iodo-4-methoxy-1-nitrobenzene
COC1=CC=C(C(I)=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 68.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 279.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Iodo-4-nitroanisole
compliance-icons compliance-icons
Product Identifier
  • 3-Iodo-4-nitroanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'