missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine
Click to view available options
Quantity:
1 g
5 g
Specifications
Quantity | 1 g |
Sensitivity | Light sensitive |
Formula Weight | 304.13 |
Chemical Name or Material | 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'