Learn More
3-Iodo-2-propynyl N-Butylcarbamate, TRC
Product Code. 30132397
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132397 5 g 5g
30136744 50 g 50g
18014020 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30132397 Supplier Toronto Research Chemicals Supplier No. TRCI7189505G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55406-53-6
Molecular Formula C8 H12 I N O2
Molecular Weight (g/mol) 281.09
Synonym Carbamic acid, butyl-, 3-iodo-2-propynyl ester (9CI), 3-Iodo-2-propynyl N-butylcarbamate, 3-Iodo-2-propynyl butylcarbamate, 3-Iodopropargyl n-butylcarbamate, Acticide IPS 20, Acticide IPW 50, Bioban I 20, Biodocarb, Biodocarb L 15BDG, Biodocarb L 20TEX, Biodocarb L 30BDG, Coatcide 123, Dekaben LMB, Denicide XR 5, Ecobois, FL 10, Fungitrol 400, Fungitrol 720, Fungitrol 820, Fungitrol 920, Fungitrol 940, Fungitrol F 10, Glycacil, Glycacil 2000, Glycasil L, Guardsan 388, Hokucide IPB, IBF 8DOP, IPBC, IPBC 20, IPBC 40, Iodocarb, Iodopropynyl butylcarbamate, MP 100, Mergal 710S, N-Butyl 3-iodo-2-propyn-1-yl carbamate, Neomid 220, Nipacide IPBC 20, Omacide IPBC, Omacide IPBC 100, P 20T, Polyphase, Polyphase 60B, Polyphase 641, Polyphase AF 1, Polyphase CST, Polyphase P 20T, Polyphase P-20T, Polyphase P100, Preventol MP 100, Preventol MP 260, Preventol MP 400, Thompson's Wood Protector, Timbertreat D, Troysan P 20T, Troysan Polyphase 60B, Troysan Polyphase AF 1, Troysan Polyphase AF 3, Troysan Polyphase P 100, Troysan Polyphase P 20T, Troysan Polyphase WD 17, Yotoru IP
IUPAC Name 3-iodoprop-2-ynyl N-butylcarbamate
SMILES CCCCNC(=O)OCC#CI
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H12INO2/c1-2-3-6-10-8(11)12-7-4-5-9/h2-3,6-7H2,1H3,(H,10,11)
Formula Weight 280.9913
Chemical Name or Material Iodocarb (IPBC)