Få mere at vide
3-Hydroxypropionic Acid, in aq solution (with varying amounts of 3,3'-Oxydipropanoic Acid), TRC
Artikelnummer. 30007334
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007334 100 mg
30007260 1 g
30007283 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007334 Leverandør Toronto Research Chemicals Leverandørnr. TRCH961903100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 503-66-2
Molekylær formel C3 H6 O3
Molekylvægt (g/mol) 90.078
Synonym Propanoic acid, 3-hydroxy-, Hydracrylic acid (6CI,7CI,8CI), 3-Hydroxypropanoic acid, 2-Deoxyglyceric acid, 3-Hydroxypropionic acid, Ethylenelactic acid, β-Hydroxypropionic acid, β-Lactic acid
IUPAC navn 3-hydroxypropanoic acid
SMIL OCCC(=O)O
Tekniske data
Kemisk navn eller materiale 3-Hydroxypropanoic Acid
Mængde 100 mg
InChI formel InChI=1S/C3H6O3/c4-2-1-3(5)6/h4H,1-2H2,(H,5,6)
Formel vægt 90.0317