Få mere at vide
3-Hydroxyoleylcarnitine (Mixture of Diastereomers), TRC
Artikelnummer. 30135117
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135117 5 mg -
30003668 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135117 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9427855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 851132-91-7
Molekylær formel C25 H47 N O5
Molekylvægt (g/mol) 441.64
Synonym (2R)-3-Carboxy-2-[[(9Z)-3-hydroxy-1-oxo-9-octadecen-1-yl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt, (2R)-3-Carboxy-2-[[(9Z)-3-hydroxy-1-oxo-9-octadecenyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt, 3-Hydroxyoctadecenoylcarnitine
IUPAC navn (3R)-3-[(Z)-3-hydroxyoctadec-9-enoyl]oxy-4-(trimethylammonio)butanoate
SMIL CCCCCCCC\C=C/CCCCCC(O)CC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C
Tekniske data
Kemisk navn eller materiale 3-Hydroxyoleylcarnitine (Mixture of Diastereomers)
Mængde 5 mg
InChI formel InChI=1S/C25H47NO5/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22(27)19-25(30)31-23(20-24(28)29)21-26(2,3)4/h12-13,22-23,27H,5-11,14-21H2,1-4H3/b13-12-/t22?,23-/m1/s1
Formel vægt 441.3454