Learn More
Alfa Aesar™ 3-Hydroxymethyl-4-nitroindole, 97%
Brand: Alfa Aesar™ H63120.MD
Additional Details : CAS Number : 124549-51-5 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
124549-51-5 | |
192.174 | |
GIZXLVHVDBXUPQ-UHFFFAOYSA-N | |
14432522 | |
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)CO |
C9H8N2O3 | |
MFCD04973980 | |
4-nitro-1h-indol-3-yl methanol, 4-nitro-1h-indol-3-yl-methanol | |
(4-nitro-1H-indol-3-yl)methanol |
Specifications
3-Hydroxymethyl-4-nitroindole | |
250mg | |
C9H8N2O3 | |
4-nitro-1h-indol-3-yl methanol, 4-nitro-1h-indol-3-yl-methanol | |
C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)CO | |
192.174 | |
192.17 | |
97% |
Undesignated | |
124549-51-5 | |
MFCD04973980 | |
GIZXLVHVDBXUPQ-UHFFFAOYSA-N | |
(4-nitro-1H-indol-3-yl)methanol | |
14432522 | |
Solid |