Få mere at vide
3-Hydroxyanthranilic Acid-d3, TRC
Artikelnummer. 30403722
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30403722 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403722 Leverandør Toronto Research Chemicals Leverandørnr. TRCH80179210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Hydroxyanthranilic Acid-d3
CAS 1794970-56-1
Renhedsgrad noter HPLC
Molekylær formel C7 2H3 H4 N O3
Mængde 10 mg
Synonym 2-Amino-4,5,6-trideuterio-3-hydroxybenzoic acid, 2-Amino-3-hydroxybenzoic-4,5,6-D3 acid, 3-Hydroxyanthranilic acid-d3
InChI formel InChI=1S/C7H7NO3/c8-6-4(7(10)11)2-1-3-5(6)9/h1-3,9H,8H2,(H,10,11)/i1D,2D,3D
SMIL [2H]c1c([2H])c(O)c(N)c(C(=O)O)c1[2H]
IUPAC navn 2-amino-4,5,6-trideuterio-3-hydroxybenzoic acid
Molekylvægt (g/mol) 156.15
Formel vægt 156.0614
Procent renhed >95