Få mere at vide
3'-Hydroxy Stanozolol-d3, TRC
Artikelnummer. 30404120
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30404120 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404120 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95353310MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3'-Hydroxystanozolol-D3
CAS 170082-17-4
Molekylær formel C21 2H3 H29 N2 O2
Mængde 10 mg
Synonym 3'-Hydroxystanozolol-D3
InChI formel InChI=1S/C21H32N2O2/c1-19-11-14-17(22-23-18(14)24)10-12(19)4-5-13-15(19)6-8-20(2)16(13)7-9-21(20,3)25/h12-13,15-16,25H,4-11H2,1-3H3,(H2,22,23,24)/t12-,13+,15-,16-,19-,20-,21-/m0/s1/i3D3
SMIL [2H]C([2H])([2H])[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]nc(O)c5C[C@]4(C)[C@H]3CC[C@]12C
Molekylvægt (g/mol) 347.51
Formel vægt 347.2652