Få mere at vide
3-Hydroxy-N,N,N-trimethylanilinium Bromide, Mikromol™
Beskrivelse
3-Hydroxy-N,N,N-trimethylanilinium Bromide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Hydroxy-N,N,N-trimethylanilinium Bromide |
| CAS | 1620-19-5 |
| Analyt- eller komponentnavne | 3-Hydroxy-N,N,N-trimethylanilinium Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H14 N O . Br |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 3-Hydroxy-N,N,N-trimethyl-benzenaminium Bromide (1:1), (m-Hydroxyphenyl)trimethylammonium bromide (6CI,7CI), Benzenaminium, 3-hydroxy-N,N,N-trimethyl-, bromide (9CI), 3-Hydroxyphenyltrimethylammonium bromide, Neostigmine Bromide Impurity A as Bromide, Neostigmine Impurity A as Bromide |
| InChI formel | InChI=1S/C9H13NO.BrH/c1-10(2,3)8-5-4-6-9(11)7-8;/h4-7H,1-3H3;1H |
| SMIL | [Br-].C[N+](C)(C)c1cccc(O)c1 |
| IUPAC navn | (3-hydroxyphenyl)-trimethylazanium;bromide |
| Vis mere |