Få mere at vide
3-Hydroxy Mepivacaine, TRC
Artikelnummer. 30136621
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136621 1 mg
30009340 5 mg
30009511 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136621 Leverandør Toronto Research Chemicals Leverandørnr. TRCH8253551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 37055-90-6
Molekylær formel C15H22N2O2
Molekylvægt (g/mol) 262.35
Synonym N-(3-Hydroxy-2,6-dimethylphenyl)-1-methyl-2-piperidinecarboxamide, 1-Methyl-3'-hydroxy-2',6'-pipecoloxylidide, 1-Methyl-4'-hydroxy-2',6'-pipecoloxylidide, 1-Methylpipecolo-3'-hydroxy-2',6'-xylidide;
SMIL CN1CCCCC1C(=O)Nc2c(C)ccc(O)c2C
Tekniske data
Kemisk navn eller materiale 3-Hydroxy Mepivacaine
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C15H22N2O2/c1-10-7-8-13(18)11(2)14(10)16-15(19)12-6-4-5-9-17(12)3/h7-8,12,18H,4-6,9H2,1-3H3,(H,16,19)
Formel vægt 262.1681
Procent renhed >95%