Få mere at vide
3-Hydroxy Detomidine-15N2,d2 Hydrochloride, TRC
Artikelnummer. 30406643
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30406643 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406643 Leverandør Toronto Research Chemicals Leverandørnr. TRCH82534210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Hydroxy Detomidine-15N2,d2 Hydrochloride
CAS 1346603-61-9
Molekylær formel C12H13D2Cl15N2O
Mængde 10 mg
Synonym 3-(1H-Imidazol-5-ylmethyl)-2-methylbenzenemethanol-15N2,d2 Hydrochloride, 3-(1H-Imidazol-4-ylmethyl)-2-methylbenzenemethanol-15N2,d2 Hydrochloride
InChI formel InChI=1S/C12H14N2O.ClH/c1-9-10(3-2-4-11(9)7-15)5-12-6-13-8-14-12;/h2-4,6,8,15H,5,7H2,1H3,(H,13,14);1H
SMIL [2H]C([2H])(C1=C(C(=CC=C1)CO)C)C2=C[15N]=C[15NH]2.Cl
Molekylvægt (g/mol) 242.71
Formel vægt 242.0939