missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Hydroxy-3-methylpentane-1,5-dioic Acid, TRC

Artikelnummer. 30137750
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137750 500 mg
30133301 1 g
30004815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30137750 Leverandør Toronto Research Chemicals Leverandørnr. TRCH948263500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 503-49-1
Molekylær formel C6 H10 O5
Molekylvægt (g/mol) 162.14
Synonym Pentanedioic acid, 3-hydroxy-3-methyl-, Glutaric acid, 3-hydroxy-3-methyl- (6CI,7CI,8CI), 3-Hydroxy-3-methylpentanedioic acid, 3-Hydroxy-3-methylglutaric acid, 3-Methyl-3-hydroxyglutaric acid, 3-Methyl-3-hydroxypentanedioic acid, CB 337, Dicrotalic acid, HMG, HMGA, Lipoglutaren, Medroglutaric acid, Meglutol, Mevalon, NSC 361411, β-Hydroxy-β-methylglutaric acid
IUPAC navn 3-hydroxy-3-methylpentanedioic acid
SMIL CC(O)(CC(=O)O)CC(=O)O

Tekniske data

Kemisk navn eller materiale 3-Hydroxy-3-methylpentane-1,5-dioic Acid
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C6H10O5/c1-6(11,2-4(7)8)3-5(9)10/h11H,2-3H2,1H3,(H,7,8)(H,9,10)
Formel vægt 162.0528
Procent renhed >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.