Få mere at vide
3-Hydroxy-3-methylglutaric-d3 Anhydride, TRC
Artikelnummer. 30404387
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30404387 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404387 Leverandør Toronto Research Chemicals Leverandørnr. TRCH948267100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Hydroxy-3-methylglutaric-d3 Anhydride
CAS 115135-38-1
Molekylær formel C6H5D3O4
Mængde 100 mg
Synonym 3-Hydroxy-3-methylpentane-1,5-dioic-d3 Anhydride, Dihydro-4-hydroxy-4-methyl-2H-pyran-2,6(3H)-dione-d3, Dicrotalic-d3 Anhydride
InChI formel InChI=1S/C6H8O4/c1-6(9)2-4(7)10-5(8)3-6/h9H,2-3H2,1H3/i1D3
SMIL [2H]C([2H])([2H])C1(O)CC(=O)OC(=O)C1
IUPAC navn 4-hydroxy-4-(trideuteriomethyl)oxane-2,6-dione
Molekylvægt (g/mol) 147.14
Formel vægt 147.0611