Få mere at vide
3-Hydroxy-2-methylbutanoic Acid Ethyl Ester(Mixture of Diastereomers), TRC
Artikelnummer. 30397841
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30397841 250 mg -
30392407 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30397841 Leverandør Toronto Research Chemicals Leverandørnr. TRCH946490250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27372-03-8
Molekylær formel C7H14O3
Molekylvægt (g/mol) 146.18
Synonym Ethyl 3-Hydroxy-2-methylbutyrate, 3-Hydroxy-2-methyl-butyric Acid Ethyl Ester
IUPAC navn ethyl 3-hydroxy-2-methylbutanoate
SMIL CCOC(=O)C(C)C(C)O
Tekniske data
Kemisk navn eller materiale 3-Hydroxy-2-methylbutanoic Acid Ethyl Ester(Mixture of Diastereomers)
Mængde 250 mg
InChI formel InChI=1S/C7H14O3/c1-4-10-7(9)5(2)6(3)8/h5-6,8H,4H2,1-3H3
Formel vægt 146.0943