Learn More
3-Hydroxy-2-methyl-d3-benzoic Acid, TRC
Product Code. 30405742
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30405742 10 mg 10mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30405742 Supplier Toronto Research Chemicals Supplier No. TRCH94640710MG
 Restricted Item
Forventet forsendelse: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Hydroxy-2-methyl-d3-benzoic Acid
CAS 1020719-51-0
Molecular Formula C8 2H3 H5 O3
Quantity 10 mg
Synonym Benzoic acid, 3-hydroxy-2-(methyl-d3)-, 3,2-Cresotic acid-d3, 3-Hydroxy-2-(methyl-d3)benzoic acid, 3-Hydroxy-2-methyl-d3-benzoic acid, 2-(Methyl-d3)-3-hydroxybenzoic acid, 3-Hydroxy-2-methylbenzoic acid D3 (methyl-D3)
InChI Formula InChI=1S/C8H8O3/c1-5-6(8(10)11)3-2-4-7(5)9/h2-4,9H,1H3,(H,10,11)/i1D3
SMILES [2H]C([2H])([2H])c1c(O)cccc1C(=O)O
IUPAC Name 3-hydroxy-2-(trideuteriomethyl)benzoic acid
Molecular Weight (g/mol) 155.17
Formula Weight 155.0662