Få mere at vide
3-Hydroxy-1-methyl-pyridinium Iodide, Mikromol™
Beskrivelse
3-Hydroxy-1-methyl-pyridinium Iodide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Hydroxy-1-methyl-pyridinium Iodide |
| CAS | 5-2-7500 |
| Analyt- eller komponentnavne | 3-Hydroxy-1-methyl-pyridinium Iodide |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C6 H8 N O . I |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 3-Hydroxy-1-methyl-pyridinium Iodide, Pyridostigmine Bromide Imp. B (EP) as Iodide, Pyridinium, 3-hydroxy-1-methyl-, iodide (1:1), 3-Hydroxy-1-methylpyridinium iodide (6CI), Pyridinium, 3-hydroxy-1-methyl-, iodide (8CI,9CI), 1-Methyl-3-hydroxypyridinium iodide, 3-Hydroxypyridinium methiodide, Pyridostigmine USP Related Compound B, Pyridostigmine USP RC B, Pyridostigmine Related Compound B as Iodide, Pyridostigmine Bromide Impurity B as Iodide, Pyridostigmine Impurity B as Iodide |
| InChI formel | InChI=1S/C6H7NO.HI/c1-7-4-2-3-6(8)5-7;/h2-5H,1H3;1H |
| SMIL | [I-].C[n+]1cccc(O)c1 |
| IUPAC navn | 1-methylpyridin-1-ium-3-ol;iodide |
| Vis mere |