Learn More
3-Guanidinopropionic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-4838143
Click to view available options
Quantity:
1 g
5 g
353-09-3
C4H9N3O2
131.13
MFCD00045939
KMXXSJLYVJEBHI-UHFFFAOYSA-N
3-guanidinopropionic acid, 3-guanidinopropanoic acid, beta-guanidinopropionic acid, guanidinopropionic acid, 3-guanidinopropanoate, 3-carbamimidamidopropanoic acid, beta-gpa, unii-ul1984yrka, beta-guanadinopropionate, n-aminoiminomethyl-beta-alanine
67701
CHEBI:15968
3-(diaminomethylideneamino)propanoic acid
C(CN=C(N)N)C(=O)O
353-09-3
C4H9N3O2
131.13
MFCD00045939
KMXXSJLYVJEBHI-UHFFFAOYSA-N
3-guanidinopropionic acid, 3-guanidinopropanoic acid, beta-guanidinopropionic acid, guanidinopropionic acid, 3-guanidinopropanoate, 3-carbamimidamidopropanoic acid, beta-gpa, unii-ul1984yrka, beta-guanadinopropionate, n-aminoiminomethyl-beta-alanine
67701
CHEBI:15968
3-(diaminomethylideneamino)propanoic acid
C(CN=C(N)N)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 222.0°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)2CO2H
Quantity 1 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material 3-Guanidinopropionic acid
compliance-icons
Product Identifier
  • 3-Guanidinopropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.