Få mere at vide
3-Fluoroaniline-2,4,6-d3,ND2, CDN
Artikelnummer. 30323555
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323555 0.5 g 0.5g
30318929 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323555 Leverandør CDN Leverandørnr. CDND74970.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Fluoroaniline-N,N,2,4,6-D5
CAS 1398065-56-9
Molekylær formel C6 D5 H F N
Mængde 0.5 g
Synonym 3-Fluoroaniline-2,4,6-d3,ND2, 3-Fluoroaniline-N,N,2,4,6-D5, 3-Fluoroaniline-d5, 3-Fluorobenzene-2,4,6-d3-amine-d2
InChI formel InChI=1 S/C6H6FN/c7-5-2-1-3-6(8)4-5/h1-4 H,8H2/i2D,3 D,4 D/hD2
SMIL [2 H]N([2 H])c1c([2 H])cc([2 H])c(F)c1[2 H]
IUPAC navn N,N,2,4,6-pentadeuterio-3-fluoroaniline
Molekylvægt (g/mol) 116.1478
Formel vægt 116.0798 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity