missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-N-methylbenzylamine, 97%

Brand:  Alfa Aesar™ H56177.06

Additional Details : CAS Number : 90389-84-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15402618

  • 1220.52 DKK / 5g


In Stock 1
Add to basket

Specifications

Chemical Identifiers

90389-84-7
139.173
ZXWCKKSSCIFVBT-UHFFFAOYSA-N
457577
CNCC1=CC(=CC=C1)F
C8H10FN
MFCD04623547
3-fluorobenzyl methylamine, n-methyl-3-fluorobenzylamine, 3-fluoro-n-methylbenzylamine, 1-3-fluorophenyl-n-methylmethanamine, benzenemethanamine, 3-fluoro-n-methyl, 3-fluorophenyl methyl methyl amine, 1-3-fluorophenyl-n-methyl-methanamine, 3-fluorophenyl-n-methylmethanamine, 3-fluorobenzyl-methylamine, acmc-209r5x
1-(3-fluorophenyl)-N-methylmethanamine

Specifications

3-Fluoro-N-methylbenzylamine
1.499
Colorless to Tan
90389-84-7
MFCD04623547
ZXWCKKSSCIFVBT-UHFFFAOYSA-N
1-(3-fluorophenyl)-N-methylmethanamine
457577
Liquid
1.015g/mL
183°C to 184°C
5g
C8H10FN
3-fluorobenzyl methylamine, n-methyl-3-fluorobenzylamine, 3-fluoro-n-methylbenzylamine, 1-3-fluorophenyl-n-methylmethanamine, benzenemethanamine, 3-fluoro-n-methyl, 3-fluorophenyl methyl methyl amine, 1-3-fluorophenyl-n-methyl-methanamine, 3-fluorophenyl-n-methylmethanamine, 3-fluorobenzyl-methylamine, acmc-209r5x
CNCC1=CC(=CC=C1)F
139.173
139.17
97%
Videos
SDS