missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™

Catalog Number 11475950
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
238754-67-1
C9H6F4O2
222.139
MFCD00236303
ZFOWDXKJQNNLMW-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl phenylacetic acid, 2-3-fluoro-4-trifluoromethyl phenyl acetic acid, 3-fluoro-4-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl phenylaceticacid, 3-fluoro-4-trifluoromethyl-phenyl-acetic acid, pubchem4186, acmc-1chko
2737591
2-[3-fluoro-4-(trifluoromethyl)phenyl]acetic acid
C1=CC(=C(C=C1CC(=O)O)F)C(F)(F)F
This item is not returnable. View return policy
238754-67-1
C9H6F4O2
222.139
MFCD00236303
ZFOWDXKJQNNLMW-UHFFFAOYSA-N
3-fluoro-4-trifluoromethyl phenylacetic acid, 2-3-fluoro-4-trifluoromethyl phenyl acetic acid, 3-fluoro-4-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-fluoro-4-trifluoromethyl, 3-fluoro-4-trifluoromethyl phenylaceticacid, 3-fluoro-4-trifluoromethyl-phenyl-acetic acid, pubchem4186, acmc-1chko
2737591
2-[3-fluoro-4-(trifluoromethyl)phenyl]acetic acid
C1=CC(=C(C=C1CC(=O)O)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 88°C to 90°C
Quantity 1 g
Formula Weight 222.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)phenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'