missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7054527
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
445303-67-3
C13H15BF4O2
290.065
MFCD09878544
FPCSARGPMNDYAO-UHFFFAOYSA-N
2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-4-trifluoromethyl phenylboronic acid pinacol ester, 3-fluoro-4-trifluoromethyl phenylboronic acid, pinacol ester, 3-fluoro-4-trifluoromethyl benzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane
46739054
2-[3-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(F)(F)F)F
This item is not returnable. View return policy
445303-67-3
C13H15BF4O2
290.065
MFCD09878544
FPCSARGPMNDYAO-UHFFFAOYSA-N
2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-4-trifluoromethyl phenylboronic acid pinacol ester, 3-fluoro-4-trifluoromethyl phenylboronic acid, pinacol ester, 3-fluoro-4-trifluoromethyl benzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane
46739054
2-[3-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(F)(F)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'