missing translation for 'onlineSavingsMsg'
Learn More
3-Fluoro-4-(trifluoromethoxy)benzaldehyde, 97% Product Code.: 11327009

3-Fluoro-4-(trifluoromethoxy)benzaldehyde, 97%

Product Code. p-7029391
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11327009

Brand: Thermo Scientific Alfa Aesar H26847.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 473917-15-6 | C8H4F4O2 | 208.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 473917-15-6
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD06660201
InChI Key RQLRUBHAAVGRPW-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethoxy benzaldehyde, 3-fluoro-4-trifluoromethoxy-benzaldehyde
PubChem CID 2783358
IUPAC Name 3-fluoro-4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)OC(F)(F)F

Specifications

Refractive Index 1.4455
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 208.11
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethoxy)benzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot NumberC26T008 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH26847.03,H26847.06
Lot Number10182004 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH26847.03,H26847.06
Lot Number10140742 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH26847.03,H26847.06
Lot Number10190974 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH26847.03,H26847.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoro-4-(trifluoromethoxy)benzaldehyde, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.