missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzoyl chloride, 97%, Thermo Scientific™

Catalog Number 15443377
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
3907-15-1
C8H6ClFO2
188.582
MFCD03428589
LOSBAHQDKBWMBY-UHFFFAOYSA-N
2779255
3-fluoro-4-methoxybenzoyl chloride
COC1=C(C=C(C=C1)C(=O)Cl)F
This item is not returnable. View return policy
3907-15-1
C8H6ClFO2
188.582
MFCD03428589
LOSBAHQDKBWMBY-UHFFFAOYSA-N
2779255
3-fluoro-4-methoxybenzoyl chloride
COC1=C(C=C(C=C1)C(=O)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 71°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Synonym 3-fluoro-4-methoxybenzoylchloride, 3-fluoro-4-methoxy-benzoyl chloride, benzoyl chloride,3-fluoro-4-methoxy, pubchem1921, acmc-20an3u, 4-chlorocarbonyl-2-fluoroanisole, 3-fluoro-4-methoxy benzoyl chloride, benzoyl chloride, 3-fluoro-4-methoxy
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methoxybenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'