missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7040041
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1408088-34-5
C17H26BFN2O2
320.215
MFCD22988988
RFECQDXNBWGCRR-UHFFFAOYSA-N
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
70974710
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F
This item is not returnable. View return policy
1408088-34-5
C17H26BFN2O2
320.215
MFCD22988988
RFECQDXNBWGCRR-UHFFFAOYSA-N
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
70974710
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 320.21
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'