missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylbenzoic acid, 98%, Thermo Scientific™

Catalog Number p-7030977
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
699-90-1
C8H7FO2
154.14
MFCD00075246
XMKZAIHFVHJGPV-UHFFFAOYSA-N
3-fluoro-2-methyl-benzoic acid, 2-methyl-3-fluoro benzoic acid, 3-fluoro-2-methylbenzoicacid, benzoic acid, 3-fluoro-2-methyl, 6-fluortoluylsaure, pubchem1329, 2-carboxy-6-fluorotoluene, acmc-209oc1, ksc352q5t, 2-methyl-3-fluorobenzoic acid
246682
3-fluoro-2-methylbenzoic acid
CC1=C(C=CC=C1F)C(=O)O
This item is not returnable. View return policy
699-90-1
C8H7FO2
154.14
MFCD00075246
XMKZAIHFVHJGPV-UHFFFAOYSA-N
3-fluoro-2-methyl-benzoic acid, 2-methyl-3-fluoro benzoic acid, 3-fluoro-2-methylbenzoicacid, benzoic acid, 3-fluoro-2-methyl, 6-fluortoluylsaure, pubchem1329, 2-carboxy-6-fluorotoluene, acmc-209oc1, ksc352q5t, 2-methyl-3-fluorobenzoic acid
246682
3-fluoro-2-methylbenzoic acid
CC1=C(C=CC=C1F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-2-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'