missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylbenzaldehyde, 98%, Thermo Scientific™

Catalog Number p-7055637
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
147624-13-3
C8H7FO
138.141
MFCD00075256
HSUCYMJBFHBMTB-UHFFFAOYSA-N
2-methyl-3-fluorobenzaldehyde, 3-fluoro-2-methyl-benzaldehyde, benzaldehyde, 3-fluoro-2-methyl, pubchem1453, acmc-1cfbd, intermediates-zcf02128, ksc494i0h, attercop-chm at111499, 3-fluoro-2-methylbenzaldehyde
2779235
3-fluoro-2-methylbenzaldehyde
CC1=C(C=CC=C1F)C=O
This item is not returnable. View return policy
147624-13-3
C8H7FO
138.141
MFCD00075256
HSUCYMJBFHBMTB-UHFFFAOYSA-N
2-methyl-3-fluorobenzaldehyde, 3-fluoro-2-methyl-benzaldehyde, benzaldehyde, 3-fluoro-2-methyl, pubchem1453, acmc-1cfbd, intermediates-zcf02128, ksc494i0h, attercop-chm at111499, 3-fluoro-2-methylbenzaldehyde
2779235
3-fluoro-2-methylbenzaldehyde
CC1=C(C=CC=C1F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.16 g/mL
Boiling Point 92°C to 93°C (20 mmHg)
Flash Point 81°C (178°F)
Refractive Index 1.526
Quantity 1 g
Beilstein 6321735
Formula Weight 138.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-2-methylbenzaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'