missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxypyridine, 97%, Thermo Scientific™

Catalog Number p-7030987
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
884494-69-3
C6H6FNO
127.12
MFCD03094945
VOXFISDIRIAWCW-UHFFFAOYSA-N
pubchem10624, 2-methoxy-fluoropyridine, 3-fluoro-2-methoxy-pyridine, 3-fluoranyl-2-methoxy-pyridine, 2-methoxy-3-fluoropyridine
44754778
3-fluoro-2-methoxypyridine
COC1=C(F)C=CC=N1
This item is not returnable. View return policy
884494-69-3
C6H6FNO
127.12
MFCD03094945
VOXFISDIRIAWCW-UHFFFAOYSA-N
pubchem10624, 2-methoxy-fluoropyridine, 3-fluoro-2-methoxy-pyridine, 3-fluoranyl-2-methoxy-pyridine, 2-methoxy-3-fluoropyridine
44754778
3-fluoro-2-methoxypyridine
COC1=C(F)C=CC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN1993
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-methoxypyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'