missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3'-Fluoro-2'-methoxyacetophenone, 97%
Brand: Alfa Aesar™ H31909.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
295779-86-1 | |
168.167 | |
XWKGHXBPAPLVPW-UHFFFAOYSA-N | |
11252225 | |
CC(=O)C1=C(C(=CC=C1)F)OC |
C9H9FO2 | |
MFCD12545933 | |
1-3-fluoro-2-methoxyphenyl ethanone, 1-3-fluoro-2-methoxyphenyl ethan-1-one, 1-3-fluoro-2-methoxy-phenyl-ethanone, 3'-fluoro-2'-methoxyacetophenone, 2'-methoxy-3'-fluoroacetophenone, ethanone, 1-3-fluoro-2-methoxyphenyl-9ci | |
1-(3-fluoro-2-methoxyphenyl)ethanone |
Specifications
3'-Fluoro-2'-methoxyacetophenone | |
295779-86-1 | |
MFCD12545933 | |
XWKGHXBPAPLVPW-UHFFFAOYSA-N | |
1-(3-fluoro-2-methoxyphenyl)ethanone | |
11252225 | |
97% |
1g | |
C9H9FO2 | |
1-3-fluoro-2-methoxyphenyl ethanone, 1-3-fluoro-2-methoxyphenyl ethan-1-one, 1-3-fluoro-2-methoxy-phenyl-ethanone, 3'-fluoro-2'-methoxyacetophenone, 2'-methoxy-3'-fluoroacetophenone, ethanone, 1-3-fluoro-2-methoxyphenyl-9ci | |
CC(=O)C1=C(C(=CC=C1)F)OC | |
168.167 | |
168.17 |