missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-2-formylbenzeneboronic acid, 95%

Brand:  Alfa Aesar™ H64909.06

Additional Details : CAS Number : 871126-15-7 Weight : 0.00500kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15472319

  • 1424.47 DKK / 5g


Estimated Shipment
25-08-2020
Add to basket

Description

Description

Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications

Chemical Identifiers

871126-15-7
167.93
GVHWLCYABLWGIR-UHFFFAOYSA-N
53412038
B(C1=C(C(=CC=C1)F)C=O)(O)O
C7H6BFO3
MFCD10697421
3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
(3-fluoro-2-formylphenyl)boronic acid

Specifications

3-Fluoro-2-formylbenzeneboronic acid
123°C to 127°C
871126-15-7
MFCD10697421
3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
GVHWLCYABLWGIR-UHFFFAOYSA-N
(3-fluoro-2-formylphenyl)boronic acid
53412038
Solid
Light sensitive
5g
C7H6BFO3
UN3261
Slightly soluble in water.
B(C1=C(C(=CC=C1)F)C=O)(O)O
167.93
167.93
95%
Videos
SDS Documents